siRNA Details
| Gene Symbol: COBLL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278458.1 | NP_001265387.1 | 15 | 2133 | ||
| NM_001278460.1 | NP_001265389.1 | 12 | 1956 | |||
| NM_001278461.1 | NP_001265390.1 | 11 | 1916 | |||
| NM_014900.4 | NP_055715.3 | 12 | 2030 | |||
| GenBank mRNA | AB023194.1 | - | 12 | 2024 | ||
| AK225849.1 | - | 12 | 1813 | |||
| AK294937.1 | - | 15 | 2133 | |||
| BC071588.1 | - | 11 | 1918 | 30345650 | ||
| BX537877.1 | - | 7 | 3518 | |||
| BX649112.1 | - | 6 | 899 | |||

COBLL1
Genomic Map