siRNA Details
| Gene Symbol: FRMPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014907.2 | NP_055722.2 | 16 | 2412 | ||
| XM_011517803.2 | XP_011516105.1 | 17 | 2814 | |||
| XM_011517804.2 | XP_011516106.1 | 16 | 2733 | |||
| XM_011517805.2 | XP_011516107.1 | 13 | 2185 | |||
| XM_011517806.2 | XP_011516108.1 | 12 | 3421 | |||
| XM_017014480.1 | XP_016869969.1 | 16 | 3093 | |||
| XM_017014481.1 | XP_016869970.1 | 12 | 2788 | |||
| XM_017014482.1 | XP_016869971.1 | 13 | 2786 | |||
| GenBank mRNA | AB023184.1 | - | 16 | 2412 | ||
| BC114965.1 | - | 16 | 2391 | 40082639 | ||
| BX648967.1 | - | 16 | 2962 | |||

FRMPD1
Genomic Map