siRNA Details
| Gene Symbol: ZNF507 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136156.1 | NP_001129628.1 | 7 | 2983 | ||
| NM_014910.4 | NP_055725.2 | 6 | 2889 | |||
| GenBank mRNA | AK126703.1 | - | 4 | 506 | ||
| AK292526.1 | - | 6 | 2887 | |||
| AL713775.1 | - | 7 | 2870 | |||
| BC110332.1 | - | 5 | 1090 | |||
| BC136417.1 | - | 8 | 2931 | 9020405 | ||
| BX640881.1 | - | 6 | 2851 | |||

ZNF507
Genomic Map