siRNA Details
| Gene Symbol: ANKRD26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256053.1 | NP_001242982.1 | 32 | 4913 | ||
| NM_014915.2 | NP_055730.2 | 32 | 4916 | |||
| XM_006717423.2 | XP_006717486.1 | 33 | 6002 | |||
| XM_006717425.3 | XP_006717488.1 | 33 | 6002 | |||
| XM_011519416.2 | XP_011517718.1 | 33 | 6002 | |||
| XM_017015928.1 | XP_016871417.1 | 33 | 6002 | |||
| XM_017015929.1 | XP_016871418.1 | 34 | 5990 | |||
| XM_017015930.1 | XP_016871419.1 | 33 | 6002 | |||
| XM_017015931.1 | XP_016871420.1 | 33 | 6002 | |||
| XM_017015932.1 | XP_016871421.1 | 33 | 6002 | |||
| XM_017015933.1 | XP_016871422.1 | 34 | 6210 | |||
| GenBank mRNA | AB028997.1 | - | 32 | 4891 | ||
| AL137351.1 | - | 5 | 734 | |||
| BC110646.1 | - | 2 | 109 | |||
| BC130453.1 | - | 32 | 4857 | 40146484 | ||

ANKRD26
Genomic Map