siRNA Details
| Gene Symbol: MORC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303256.2 | NP_001290185.1 | 9, 10 | 1228 | ||
| NM_001303257.2 | NP_001290186.1 | 9, 10 | 1228 | |||
| NM_014941.3 | NP_055756.1 | 10, 11 | 2094 | |||
| XM_011530004.1 | XP_011528306.1 | 9, 10 | 1453 | |||
| XM_017028667.1 | XP_016884156.1 | 9, 10 | 1453 | |||
| XM_017028668.1 | XP_016884157.1 | 10, 11 | 1548 | |||
| GenBank mRNA | AB020659.1 | - | 10, 11 | 2008 | ||
| BC019257.1 | - | 8, 9 | 755 | 3503651 | ||
| BC136782.1 | - | 10, 11 | 953 | 9020772 | ||
| CR456469.1 | - | 8, 9 | 754 | |||

MORC2
Genomic Map