siRNA Details
| Gene Symbol: KIAA0907 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014949.3 | NP_055764.2 | 10 | 1289 | ||
| GenBank mRNA | AB020714.1 | - | 10 | 1246 | ||
| AK289902.1 | - | 10 | 1128 | |||
| AY112680.1 | - | 10 | 1220 | |||
| AY112681.1 | - | 10 | 1220 | |||
| AY112682.1 | - | 10 | 1119 | |||
| BC062637.1 | - | 10 | 1244 | 5497581 | ||

KIAA0907
Genomic Map