siRNA Details
| Gene Symbol: ZBTB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001123329.1 | NP_001116801.1 | 2 | 2168 | ||
| NM_014950.2 | NP_055765.2 | 2 | 2168 | |||
| XM_005267410.1 | XP_005267467.1 | 4 | 2304 | |||
| XM_011536566.2 | XP_011534868.1 | 3 | 2041 | |||
| XM_011536568.2 | XP_011534870.1 | 4 | 2304 | |||
| XM_017021094.1 | XP_016876583.1 | 4 | 3180 | |||
| XM_017021095.1 | XP_016876584.1 | 3 | 2096 | |||
| GenBank mRNA | AB023214.1 | - | 2 | 2039 | ||
| AK291743.1 | - | 2 | 1946 | |||
| AY598321.1 | - | 1 | 1777 | |||
| BC050719.1 | - | 4 | 2208 | 6141266 | ||

ZBTB1
Genomic Map