siRNA Details
| Gene Symbol: RPH3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143854.1 | NP_001137326.1 | 5 | 589 | ||
| NM_014954.3 | NP_055769.2 | 4 | 577 | |||
| XM_011538048.1 | XP_011536350.1 | 5 | 393 | |||
| GenBank mRNA | AB023202.1 | - | 5 | 510 | ||
| AK295696.1 | - | 5 | 543 | |||
| AK316112.1 | - | 5 | 424 | |||
| BC002605.2 | - | 3 | 315 | |||
| BC009843.2 | - | 3 | 238 | |||
| BC017259.2 | - | 4 | 384 | 3510158 | ||

RPH3A
Genomic Map