siRNA Details
| Gene Symbol: RPIA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144563.2 | NP_653164.2 | 2 | 342 | ||
| GenBank mRNA | AY050633.1 | - | 2 | 318 | ||
| BC015529.1 | - | 2 | 314 | 3904718 | ||
| CB989690.1 | - | 2 | 376 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_009075, L35034, AK166785, BC053526, AK137235.

RPIA
Genomic Map