siRNA Details
| Gene Symbol: KIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012311.3 | NP_036443.1 | 13 | 1236 | ||
| XM_017015946.1 | XP_016871435.1 | 12 | 1107 | |||
| GenBank mRNA | AJ005273.1 | - | 13 | 1207 | ||
| BC017309.2 | - | 13 | 1153 | 5089578 | ||

KIN
Genomic Map