siRNA Details
| Gene Symbol: KDM2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012308.2 | NP_036440.1 | 11 | 1912 | ||
| NR_027473.1 | 11 | 1912 | ||||
| XM_006718480.3 | XP_006718543.1 | 7 | 930 | |||
| XM_011544860.2 | XP_011543162.1 | 11 | 1971 | |||
| XM_011544861.1 | XP_011543163.1 | 6 | 624 | |||
| XM_017017391.1 | XP_016872880.1 | 11 | 1218 | |||
| XM_017017392.1 | XP_016872881.1 | 10 | 1029 | |||
| GenBank mRNA | JQ710744.1 | - | 12 | 1265 | ||

KDM2A
Genomic Map