siRNA Details
Gene Symbol: CEP152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001194998.1 | NP_001181927.1 | 14 | 2053 | ||
NM_014985.3 | NP_055800.2 | 14 | 2053 | |||
XM_006720437.3 | XP_006720500.1 | 14 | 2238 | |||
XM_011521373.2 | XP_011519675.1 | 14 | 2806 | |||
XM_011521374.2 | XP_011519676.1 | 14 | 2805 | |||
XM_011521375.2 | XP_011519677.1 | 14 | 2805 | |||
XM_011521378.2 | XP_011519680.1 | 14 | 2805 | |||
XM_011521379.2 | XP_011519681.1 | 14 | 2804 | |||
XM_011521381.2 | XP_011519683.1 | 2 | 966 | |||
XM_017022014.1 | XP_016877503.1 | 7 | 999 | |||
XM_017022015.1 | XP_016877504.1 | 2 | 155 | |||
XM_017022016.1 | XP_016877505.1 | 14 | 2808 | |||
GenBank mRNA | AB020719.1 | - | 13 | 1658 | ||
AK303805.1 | - | 14 | 1976 | |||
BC117182.1 | - | 14 | 1894 | 40125733 | ||
BX648822.1 | - | 14 | 1980 |
