siRNA Details
| Gene Symbol: KLHDC10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014997.3 | NP_055812.1 | 4, 5 | 756 | ||
| XM_005250233.4 | XP_005250290.1 | 3, 4 | 645 | |||
| GenBank mRNA | BC009977.2 | - | 2, 3 | 169 | ||
| BC036464.2 | - | 4, 5 | 738 | 4828273 | ||
| BC044884.1 | - | 4, 5 | 758 | 5273087 | ||
| D87454.1 | - | 4, 5 | 498 | |||

KLHDC10
Genomic Map