siRNA Details
| Gene Symbol: SAMD4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161576.2 | NP_001155048.2 | 2 | 596 | ||
| NM_015589.5 | NP_056404.4 | 2 | 596 | |||
| GenBank mRNA | AF429970.1 | - | 2 | 399 | ||
| AK024652.1 | - | 2 | 289 | |||
| BC057838.1 | - | 2 | 577 | |||
| BC121173.1 | - | 2 | 293 | 40122465 | ||
| BC121174.1 | - | 2 | 293 | 40122467 | ||
| BX648198.1 | - | 6 | 3015 | |||

SAMD4A
Genomic Map