siRNA Details
| Gene Symbol: GGA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015044.4 | NP_055859.1 | 17 | 1880 | ||
| XM_017023074.1 | XP_016878563.1 | 18 | 5569 | |||
| XM_017023075.1 | XP_016878564.1 | 17 | 1883 | |||
| GenBank mRNA | AB029003.1 | - | 14 | 1510 | ||
| AF165531.1 | - | 17 | 1818 | |||
| AF190863.1 | - | 17 | 1824 | |||
| AF233522.1 | - | 17 | 1823 | |||
| AF323754.1 | - | 18 | 1736 | |||
| AK024619.1 | - | 17 | 1879 | |||
| AK093769.1 | - | 2 | 763 | |||
| BC000284.1 | - | 17 | 1862 | 3356490 | ||

GGA2
Genomic Map