siRNA Details
| Gene Symbol: EMC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271427.1 | NP_001258356.1 | 15 | 1757 | ||
| NM_001271428.1 | NP_001258357.1 | 15 | 1757 | |||
| NM_001271429.1 | NP_001258358.1 | 14 | 1694 | |||
| NM_015047.2 | NP_055862.1 | 15 | 1760 | |||
| XM_005245787.1 | XP_005245844.1 | 16 | 1764 | |||
| XM_005245788.1 | XP_005245845.1 | 16 | 1761 | |||
| GenBank mRNA | AK025853.1 | - | 2 | 121 | ||
| AK291618.1 | - | 15 | 1723 | |||
| BC034589.1 | - | 15 | 1753 | 4831005 | ||
| BX648627.1 | - | 15 | 1739 | |||
| BX648708.1 | - | 15 | 1715 | |||
| D42044.2 | - | 15 | 1704 | |||

EMC1
Genomic Map