siRNA Details
| Gene Symbol: SWAP70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297714.1 | NP_001284643.1 | 7 | 1066 | ||
| NM_015055.3 | NP_055870.2 | 8 | 1240 | |||
| XM_005252830.4 | XP_005252887.1 | 6 | 1141 | |||
| GenBank mRNA | AB014540.1 | - | 8 | 1192 | ||
| AF134894.1 | - | 8 | 1211 | |||
| AF161439.1 | - | 2 | 77 | |||
| AF210818.1 | - | 8 | 1137 | |||
| AK027131.1 | - | 5 | 667 | |||
| AK096859.1 | - | 7 | 1066 | |||
| AK304104.1 | - | 8 | 1104 | |||
| BC000616.2 | - | 8 | 1213 | 3346240 | ||

SWAP70
Genomic Map