siRNA Details
| Gene Symbol: FAM179B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308120.1 | NP_001295049.1 | 1 | 2166 | ||
| NM_015091.3 | NP_055906.2 | 1 | 2166 | |||
| NR_131765.1 | 1 | 2166 | ||||
| XM_011536571.1 | XP_011534873.1 | 1 | 2137 | |||
| XM_017021098.1 | XP_016876587.1 | 1 | 2137 | |||
| XM_017021099.1 | XP_016876588.1 | 1 | 2137 | |||
| XM_017021100.1 | XP_016876589.1 | 1 | 2137 | |||
| GenBank mRNA | AB007883.2 | - | 1 | 2148 | ||
| AK096934.1 | - | 1 | 2137 | |||
| AK295185.1 | - | 1 | 2166 | |||
| AK307722.1 | - | 2 | 1963 | |||
| AK311034.1 | - | 1 | 2137 | |||
| BC057255.2 | - | 1 | 2143 | 5274847 | ||
| BX648723.1 | - | 1 | 2117 | |||
| CR749557.1 | - | 1 | 2144 | |||

FAM179B
Genomic Map