siRNA Details
| Gene Symbol: HIC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015094.2 | NP_055909.2 | 3 | 1101 | ||
| XM_011530007.2 | XP_011528309.1 | 3 | 3375 | |||
| XM_011530008.2 | XP_011528310.1 | 3 | 1291 | |||
| XM_011530009.2 | XP_011528311.1 | 4 | 2737 | |||
| XM_011530010.2 | XP_011528312.1 | 3 | 1287 | |||
| XM_017028669.1 | XP_016884158.1 | 2 | 1004 | |||
| GenBank mRNA | AB028943.2 | - | 3 | 1101 | ||
| AF349035.2 | - | 1 | 845 | |||
| BC094787.1 | - | 3 | 1034 | 30531041 | ||
| CR456377.1 | - | 2 | 977 | |||

HIC2
Genomic Map