siRNA Details
| Gene Symbol: PLXND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015103.2 | NP_055918.2 | 11 | 2572 | ||
| XM_011512588.1 | XP_011510890.1 | 11 | 2487 | |||
| XM_011512589.1 | XP_011510891.1 | 8 | 2097 | |||
| XM_011512590.2 | XP_011510892.1 | 11 | 2487 | |||
| GenBank mRNA | AB014520.2 | - | 11 | 2651 | ||
| AY116661.1 | - | 11 | 2652 | |||
| BC150280.1 | - | 11 | 2651 | 9007263 | ||

PLXND1
Genomic Map