siRNA Details
| Gene Symbol: PLXND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015103.2 | NP_055918.2 | 24 | 4334 | ||
| XM_011512588.1 | XP_011510890.1 | 24 | 4249 | |||
| XM_011512589.1 | XP_011510891.1 | 21 | 3859 | |||
| XM_011512590.2 | XP_011510892.1 | 24 | 4249 | |||
| XM_011512592.1 | XP_011510894.1 | 12 | 1417 | |||
| GenBank mRNA | AB014520.2 | - | 24 | 4413 | ||
| AK299425.1 | - | 3 | 166 | |||
| AY116661.1 | - | 24 | 4414 | |||
| BC003526.1 | - | 2 | 81 | |||
| BC011848.2 | - | 1 | 17 | |||
| BC150280.1 | - | 24 | 4413 | 9007263 | ||

PLXND1
Genomic Map