siRNA Details
| Gene Symbol: GSE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134473.2 | NP_001127945.1 | 12 | 2844 | ||
| NM_001278184.1 | NP_001265113.1 | 12 | 2898 | |||
| NM_014615.3 | NP_055430.1 | 13 | 3117 | |||
| XM_005255859.4 | XP_005255916.3 | 14 | 5259 | |||
| XM_005255860.3 | XP_005255917.3 | 13 | 5152 | |||
| XM_005255861.4 | XP_005255918.3 | 13 | 5040 | |||
| XM_005255863.4 | XP_005255920.1 | 13 | 3009 | |||
| XM_005255864.4 | XP_005255921.1 | 13 | 3741 | |||
| XM_005255865.4 | XP_005255922.1 | 12 | 3522 | |||
| XM_011522965.2 | XP_011521267.1 | 13 | 3508 | |||
| XM_011522966.2 | XP_011521268.1 | 13 | 2975 | |||
| XM_017023083.1 | XP_016878572.1 | 13 | 3267 | |||
| GenBank mRNA | AK127344.1 | - | 2 | 1528 | ||
| AK308612.1 | - | 3 | 566 | |||
| BC000753.2 | - | 1 | 86 | |||
| BC037556.1 | - | 12 | 2867 | 4994198 | ||
| D80004.2 | - | 12 | 2782 | |||

GSE1
Genomic Map