siRNA Details
| Gene Symbol: RRS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015169.3 | NP_055984.1 | 1 | 644 | ||
| GenBank mRNA | AK024716.1 | - | 1 | 634 | ||
| BC001811.2 | - | 1 | 629 | 2824034 | ||
| BC013043.2 | - | 1 | 639 | 3603972 | ||
| D25218.1 | - | 1 | 644 | |||

RRS1
Genomic Map