siRNA Details
| Gene Symbol: RHOBTB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160036.1 | NP_001153508.1 | 8, 9 | 1977 | ||
| NM_001160037.1 | NP_001153509.1 | 6, 7 | 1673 | |||
| NM_015178.2 | NP_055993.2 | 6, 7 | 2165 | |||
| XM_017013251.1 | XP_016868740.1 | 7, 8 | 6492 | |||
| GenBank mRNA | AB018260.2 | - | 6, 7 | 2165 | ||
| AK292863.1 | - | 6, 7 | 1813 | |||
| AK314366.1 | - | 6, 7 | 1928 | |||
| AY009093.1 | - | 6, 7 | 1819 | |||
| BC034917.1 | - | 8, 9 | 1977 | 4647716 | ||

RHOBTB2
Genomic Map