siRNA Details
| Gene Symbol: NUP210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024923.3 | NP_079199.2 | 33 | 4690 | ||
| XM_017006018.1 | XP_016861507.1 | 15 | 2523 | |||
| XM_017006019.1 | XP_016861508.1 | 15 | 2490 | |||
| XM_017006020.1 | XP_016861509.1 | 15 | 4562 | |||
| GenBank mRNA | AB020713.1 | - | 13 | 1706 | ||
| AK074101.1 | - | 18 | 2312 | |||
| AK075545.1 | - | 3 | 385 | |||
| AL117527.1 | - | 2 | 237 | |||

NUP210
Genomic Map