siRNA Details
| Gene Symbol: IQCE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287499.1 | NP_001274428.1 | 6 | 623 | ||
| NM_001287500.1 | NP_001274429.1 | 5 | 575 | |||
| NM_001287501.1 | NP_001274430.1 | 4 | 529 | |||
| NM_001287502.1 | NP_001274431.1 | 4 | 529 | |||
| NM_152558.4 | NP_689771.3 | 6 | 623 | |||
| XM_006715676.2 | XP_006715739.1 | 4 | 377 | |||
| XM_011515242.1 | XP_011513544.1 | 5 | 482 | |||
| XM_011515243.1 | XP_011513545.1 | 4 | 504 | |||
| XM_017011902.1 | XP_016867391.1 | 6 | 685 | |||
| XM_017011903.1 | XP_016867392.1 | 6 | 563 | |||
| GenBank mRNA | AK092404.1 | - | 6 | 517 | ||
| AK293372.1 | - | 5 | 398 | |||
| AK302053.1 | - | 4 | 477 | |||
| AL136792.1 | - | 5 | 304 | |||
| BC043150.1 | - | 4 | 449 | 5297871 | ||
| BC061518.1 | - | 5 | 459 | |||
| BC071858.1 | - | 6 | 597 | 5478116 | ||
| KJ534872.1 | - | 6 | 413 | |||
| KJ535081.1 | - | 4 | 319 | |||

IQCE
Genomic Map