siRNA Details
| Gene Symbol: NCAPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015261.2 | NP_056076.1 | 18 | 2904 | ||
| XM_011542719.2 | XP_011541021.1 | 17 | 2165 | |||
| GenBank mRNA | AK025667.1 | - | 2 | 195 | ||
| AK124878.1 | - | 18 | 2904 | |||
| BC098398.1 | - | 18 | 2363 | 30528692 | ||
| BC144524.1 | - | 18 | 2588 | |||
| D29954.1 | - | 18 | 2326 | |||
 Back To Top
 Back To Top
 NCAPD3
NCAPD3 Genomic Map
Genomic Map Back To Top
 Back To Top