siRNA Details
| Gene Symbol: DMXL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001174116.1 | NP_001167587.1 | 27, 28 | 7194 | ||
| NM_001174117.1 | NP_001167588.1 | 25, 26 | 5283 | |||
| NM_015263.3 | NP_056078.2 | 27, 28 | 7191 | |||
| XM_005254255.1 | XP_005254312.1 | 27, 28 | 7257 | |||
| XM_005254256.1 | XP_005254313.1 | 27, 28 | 7254 | |||
| XM_017022034.1 | XP_016877523.1 | 27, 28 | 6930 | |||
| GenBank mRNA | AB020663.2 | - | 15, 16 | 4571 | ||
| AF389880.1 | - | 27, 28 | 7173 | |||
| BC016032.1 | - | 4, 5 | 1343 | |||
| BC140781.1 | - | 25, 26 | 5148 | 9021649 | ||
| BC144539.1 | - | 27, 28 | 7059 | 9053066 | ||

DMXL2
Genomic Map