siRNA Details
| Gene Symbol: SLC9A8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001260491.1 | NP_001247420.1 | 5 | 616 | ||
| NM_015266.2 | NP_056081.1 | 5 | 616 | |||
| NR_048537.1 | 5 | 616 | ||||
| NR_048539.1 | 5 | 616 | ||||
| NR_048540.1 | 5 | 616 | ||||
| XM_006723756.1 | XP_006723819.1 | 5 | 478 | |||
| XM_011528736.1 | XP_011527038.1 | 5 | 478 | |||
| XM_011528738.1 | XP_011527040.1 | 5 | 478 | |||
| XM_011528739.1 | XP_011527041.1 | 5 | 478 | |||
| XM_011528740.1 | XP_011527042.1 | 5 | 478 | |||
| XM_011528741.1 | XP_011527043.1 | 5 | 478 | |||
| XM_011528745.1 | XP_011527047.1 | 5 | 478 | |||
| XM_017027754.1 | XP_016883243.1 | 5 | 478 | |||
| XM_017027755.1 | XP_016883244.1 | 5 | 422 | |||
| XM_017027756.1 | XP_016883245.1 | 5 | 422 | |||
| GenBank mRNA | AB023156.2 | - | 5 | 448 | ||
| AK131001.1 | - | 5 | 418 | |||
| AK295803.1 | - | 6 | 419 | |||
| AK295822.1 | - | 5 | 409 | |||
| AK300318.1 | - | 5 | 501 | |||
| AK308328.1 | - | 5 | 419 | |||
| AK310530.1 | - | 5 | 418 | |||
| BC112213.1 | - | 5 | 438 | 8327681 | ||
| CR749638.1 | - | 5 | 493 | |||

SLC9A8
Genomic Map