siRNA Details
| Gene Symbol: SASH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015278.3 | NP_056093.3 | 11 | 1730 | ||
| XM_005266875.1 | XP_005266932.1 | 4 | 956 | |||
| XM_011535640.1 | XP_011533942.1 | 11 | 1173 | |||
| XM_017010598.1 | XP_016866087.1 | 11 | 2451 | |||
| XM_017010599.1 | XP_016866088.1 | 11 | 1774 | |||
| XM_017010600.1 | XP_016866089.1 | 11 | 1662 | |||
| XM_017010601.1 | XP_016866090.1 | 10 | 1876 | |||
| XM_017010602.1 | XP_016866091.1 | 9 | 1573 | |||
| XM_017010603.1 | XP_016866092.1 | 9 | 1512 | |||
| XM_017010604.1 | XP_016866093.1 | 8 | 1421 | |||
| XM_017010605.1 | XP_016866094.1 | 8 | 1387 | |||
| XM_017010606.1 | XP_016866095.1 | 4 | 1445 | |||
| XM_017010607.1 | XP_016866096.1 | 12 | 1353 | |||
| GenBank mRNA | AB018333.1 | - | 11 | 1473 | ||
| AJ507735.1 | - | 11 | 1261 | |||
| AK024403.1 | - | 5 | 715 | |||
| AK025495.1 | - | 11 | 1730 | |||
| AK074249.1 | - | 1 | 15 | |||
| AK300403.1 | - | 4 | 640 | |||
| BC063279.1 | - | 11 | 1199 | |||
| BX537611.1 | - | 1 | 924 | |||

SASH1
Genomic Map