siRNA Details
| Gene Symbol: WDR7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015285.2 | NP_056100.2 | 15 | 2768 | ||
| NM_052834.2 | NP_443066.2 | 15 | 2768 | |||
| XM_006722431.2 | XP_006722494.1 | 15 | 2700 | |||
| XM_011525888.1 | XP_011524190.1 | 15 | 2810 | |||
| XM_017025682.1 | XP_016881171.1 | 15 | 2673 | |||
| XM_017025683.1 | XP_016881172.1 | 5 | 1161 | |||
| GenBank mRNA | AB011113.2 | - | 15 | 2731 | ||
| AF188125.1 | - | 14 | 2557 | |||
| AK057141.1 | - | 15 | 2649 | |||
| AY099325.1 | - | 14 | 2557 | |||
| BC131558.1 | - | 3 | 532 | 40108655 | ||
| BC150282.1 | - | 15 | 2731 | 9007268 | ||

WDR7
Genomic Map