siRNA Details
| Gene Symbol: DNAJC16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287811.1 | NP_001274740.1 | 8 | 1233 | ||
| NM_015291.3 | NP_056106.1 | 9 | 1418 | |||
| NR_109898.1 | 9 | 1418 | ||||
| GenBank mRNA | AB023179.2 | - | 9 | 1374 | ||
| AK023805.1 | - | 5 | 588 | |||
| AK091848.1 | - | 9 | 1376 | |||
| AK292479.1 | - | 9 | 1350 | |||
| AL137715.1 | - | 3 | 348 | |||
| AL833299.1 | - | 9 | 1357 | |||
| BC047363.1 | - | 9 | 1418 | 5269539 | ||
| BX648982.1 | - | 9 | 1343 | |||
| CR749259.1 | - | 8 | 1178 | |||
| CR749567.1 | - | 8 | 1178 | |||

DNAJC16
Genomic Map