siRNA Details
| Gene Symbol: NCSTN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290184.1 | NP_001277113.1 | 15 | 1858 | ||
| NM_001290186.1 | NP_001277115.1 | 11 | 1305 | |||
| NM_015331.2 | NP_056146.1 | 14 | 1719 | |||
| XM_011509363.2 | XP_011507665.1 | 14 | 1936 | |||
| GenBank mRNA | AF240468.1 | - | 14 | 1737 | ||
| AK296153.1 | - | 9 | 1004 | |||
| AK299142.1 | - | 11 | 1192 | |||
| AK314764.1 | - | 14 | 1626 | |||
| AY359120.1 | - | 14 | 1596 | |||
| BC047621.1 | - | 15 | 1815 | 4827966 | ||
| D87442.1 | - | 14 | 1593 | |||

NCSTN
Genomic Map