siRNA Details
| Gene Symbol: MED13L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015335.4 | NP_056150.1 | 14, 15 | 2628 | ||
| XM_011538080.2 | XP_011536382.1 | 14, 15 | 2661 | |||
| XM_011538082.2 | XP_011536384.1 | 14, 15 | 2831 | |||
| GenBank mRNA | AB028948.2 | - | 8, 9 | 1695 | ||
| AF515599.1 | - | 14, 15 | 2628 | |||
| AK127706.1 | - | 11, 12 | 2162 | |||
| AL133033.1 | - | 8, 9 | 1668 | |||
| AY338463.1 | - | 14, 15 | 2780 | |||
| BC130422.1 | - | 14, 15 | 2624 | 40146453 | ||

MED13L
Genomic Map