siRNA Details
| Gene Symbol: PIP5K1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012398.2 | NP_036530.1 | 16, 17 | 2006 | ||
| XM_017026540.1 | XP_016882029.1 | 15, 16 | 1980 | |||
| GenBank mRNA | AB011161.1 | - | 16, 17 | 1974 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PIP5K1C| Gene Symbol: PIP5K1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012398.2 | NP_036530.1 | 16, 17 | 2006 | ||
| XM_017026540.1 | XP_016882029.1 | 15, 16 | 1980 | |||
| GenBank mRNA | AB011161.1 | - | 16, 17 | 1974 | ||