siRNA Details
| Gene Symbol: ICMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012405.3 | NP_036537.1 | 4, 5 | 736 | ||
| XM_011541140.2 | XP_011539442.1 | 3, 4 | 436 | |||
| GenBank mRNA | AF064084.1 | - | 4, 5 | 706 | ||
| AK092890.1 | - | 5, 6 | 677 | |||
| BC017037.1 | - | 5, 6 | 831 | |||
| BC028168.1 | - | 4, 5 | 692 | 5245125 | ||
| CR541671.1 | - | 4, 5 | 663 | |||
| CR541711.1 | - | 4, 5 | 663 | |||

ICMT
Genomic Map