siRNA Details
| Gene Symbol: GLTSCR1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318819.1 | NP_001305748.1 | 7 | 1818 | ||
| NM_015349.2 | NP_056164.1 | 5 | 1551 | |||
| GenBank mRNA | AK091434.1 | - | 5 | 1443 | ||
| AK093526.1 | - | 5 | 2162 | |||
| AK294706.1 | - | 5 | 1443 | |||
| AL833540.1 | - | 5 | 1551 | |||
| BC130302.1 | - | 5 | 1474 | 40146333 | ||
| CR627378.1 | - | 6 | 1516 | |||
| D87077.2 | - | 8 | 1715 | |||

GLTSCR1L
Genomic Map