siRNA Details
| Gene Symbol: MORC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320445.1 | NP_001307374.1 | 3 | 402 | ||
| NM_001320446.1 | NP_001307375.1 | 5 | 561 | |||
| NM_015358.2 | NP_056173.1 | 4 | 475 | |||
| XM_011529518.2 | XP_011527820.1 | 4 | 695 | |||
| XM_017028311.1 | XP_016883800.1 | 4 | 497 | |||
| XM_017028312.1 | XP_016883801.1 | 4 | 418 | |||
| GenBank mRNA | AK292957.1 | - | 4 | 426 | ||
| AK295133.1 | - | 3 | 394 | |||
| BC007311.1 | - | 4 | 434 | |||
| BC058002.1 | - | 4 | 479 | |||
| BC094779.1 | - | 4 | 432 | 30527767 | ||
| BC132731.1 | - | 4 | 429 | 40146753 | ||
| BC144116.1 | - | 5 | 515 | |||
| BU657895.1 | - | 4 | 466 | |||
| D50926.2 | - | 4 | 434 | |||
| DA434361.1 | - | 4 | 475 | |||

MORC3
Genomic Map