siRNA Details
| Gene Symbol: TSPAN15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012339.3 | NP_036471.1 | 2, 3 | 395 | ||
| XM_017016010.1 | XP_016871499.1 | 2, 3 | 401 | |||
| GenBank mRNA | AF120266.1 | - | 2, 3 | 386 | ||
| BC003157.1 | - | 2, 3 | 389 | 2958221 | ||
| BC004161.2 | - | 2, 3 | 389 | 2958221 | ||

TSPAN15
Genomic Map