siRNA Details
| Gene Symbol: PADI4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012387.2 | NP_036519.2 | 2, 3 | 301 | ||
| XM_011541150.1 | XP_011539452.1 | 2, 3 | 299 | |||
| XM_011541151.1 | XP_011539453.1 | 2, 3 | 301 | |||
| XM_011541153.1 | XP_011539455.1 | 2, 3 | 301 | |||
| XM_011541154.2 | XP_011539456.1 | 2, 3 | 301 | |||
| XM_011541155.1 | XP_011539457.1 | 2, 3 | 301 | |||
| XM_011541156.1 | XP_011539458.1 | 2, 3 | 301 | |||
| GenBank mRNA | AB017919.1 | - | 2, 3 | 299 | ||
| AF229067.1 | - | 2, 3 | 302 | |||
| AK309637.1 | - | 2, 3 | 293 | |||
| AK314839.1 | - | 2, 3 | 301 | |||
| BC025718.1 | - | 2, 3 | 285 | 5228777 | ||

PADI4
Genomic Map