siRNA Details
| Gene Symbol: DDX58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014314.3 | NP_055129.2 | 5 | 828 | ||
| GenBank mRNA | AF038963.1 | - | 5 | 827 | ||
| AK125989.1 | - | 6 | 1033 | |||
| AK301678.1 | - | 4 | 551 | |||
| AK315040.1 | - | 5 | 699 | |||
| BC107731.1 | - | 5 | 850 | |||
| BC132786.1 | - | 5 | 776 | 40146808 | ||
| BC136610.1 | - | 5 | 776 | 9020600 | ||
| BX647917.1 | - | 4 | 565 | |||

DDX58
Genomic Map