siRNA Details
| Gene Symbol: PDSS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321978.1 | NP_001308907.1 | 10 | 1320 | ||
| NM_001321979.1 | NP_001308908.1 | 12 | 1594 | |||
| NM_014317.4 | NP_055132.2 | 12 | 1511 | |||
| XM_011519437.2 | XP_011517739.1 | 9 | 1911 | |||
| XM_017016011.1 | XP_016871500.1 | 10 | 989 | |||
| GenBank mRNA | AB209763.1 | - | 11 | 3553 | ||
| AB210838.1 | - | 12 | 1269 | |||
| AF118395.1 | - | 11 | 1131 | |||
| AK024802.1 | - | 12 | 1276 | |||
| AK223414.1 | - | 12 | 1276 | |||
| AK296288.1 | - | 5 | 581 | |||
| BC049211.1 | - | 12 | 1221 | |||
| BC063635.1 | - | 10 | 1048 | 4397753 | ||

PDSS1
Genomic Map