siRNA Details
| Gene Symbol: PDSS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321979.1 | NP_001308908.1 | 10 | 1300 | ||
| NM_014317.4 | NP_055132.2 | 10 | 1217 | |||
| XM_011519437.2 | XP_011517739.1 | 7 | 1617 | |||
| XM_017016011.1 | XP_016871500.1 | 8 | 695 | |||
| GenBank mRNA | AB209763.1 | - | 9 | 3259 | ||
| AB210838.1 | - | 10 | 975 | |||
| AF118395.1 | - | 9 | 837 | |||
| AK024802.1 | - | 10 | 982 | |||
| AK223414.1 | - | 10 | 982 | |||
| BC049211.1 | - | 10 | 927 | |||

PDSS1
Genomic Map