siRNA Details
Gene Symbol: SSBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256732.1 | NP_001243661.1 | 15 | 1176 | ||
NM_001256733.1 | NP_001243662.1 | 14 | 1092 | |||
NM_001256734.1 | NP_001243663.1 | 14 | 1086 | |||
NM_001256735.1 | NP_001243664.1 | 14 | 1062 | |||
NM_001256736.1 | NP_001243665.1 | 14 | 1062 | |||
NM_012446.3 | NP_036578.2 | 15 | 1152 | |||
XM_011543297.2 | XP_011541599.1 | 15 | 1091 | |||
XM_017009304.1 | XP_016864793.1 | 15 | 994 | |||
XM_017009305.1 | XP_016864794.1 | 16 | 1083 | |||
XM_017009306.1 | XP_016864795.1 | 15 | 1067 | |||
XM_017009307.1 | XP_016864796.1 | 14 | 880 | |||
XM_017009308.1 | XP_016864797.1 | 16 | 1255 | |||
XM_017009309.1 | XP_016864798.1 | 15 | 1319 | |||
XM_017009310.1 | XP_016864799.1 | 15 | 1467 | |||
GenBank mRNA | AF077048.1 | - | 15 | 1016 | ||
AF161465.1 | - | 15 | 982 | |||
AK074537.1 | - | 15 | 947 | |||
AK293544.1 | - | 14 | 887 | |||
AK294864.1 | - | 14 | 857 | |||
AK297494.1 | - | 15 | 860 | |||
AK299858.1 | - | 14 | 930 | |||
AK312337.1 | - | 15 | 996 | |||
AL080076.1 | - | 15 | 1000 | |||
BC017020.1 | - | 15 | 1000 | 3913337 | ||
BX397789.2 | - | 3 | 743 | |||
CR936787.1 | - | 14 | 907 | |||
DA202987.1 | - | 7 | 342 |
