siRNA Details
| Gene Symbol: LSM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF182291.1 | - | 1, 2 | 56 | ||
| AJ238097.1 | - | 1, 2 | 49 | |||
| AK022772.1 | - | 1, 2 | 69 | |||
| AK024217.1 | - | 1, 2 | 78 | |||
| BC005938.1 | - | 1, 2 | 55 | 4132686 | ||
| BF667238.1 | - | 1, 2 | 96 | |||

LSM5
Genomic Map