siRNA Details
| Gene Symbol: SH3BP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014521.2 | NP_055336.1 | 4 | 1582 | ||
| XM_011510891.2 | XP_011509193.1 | 4 | 1602 | |||
| XM_011510892.1 | XP_011509194.1 | 4 | 1591 | |||
| XM_011510893.1 | XP_011509195.1 | 4 | 1516 | |||
| XM_011510894.2 | XP_011509196.1 | 4 | 4015 | |||
| GenBank mRNA | AF015043.1 | - | 1 | 970 | ||
| AF147747.1 | - | 4 | 1524 | |||
| AK291209.1 | - | 4 | 1549 | |||
| BC057396.1 | - | 4 | 1593 | 6138465 | ||
| BX647198.1 | - | 4 | 1509 | |||
| BX648877.1 | - | 4 | 1697 | |||
| DQ232895.1 | - | 4 | 1556 | |||

SH3BP4
Genomic Map