siRNA Details
| Gene Symbol: FLRT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013281.3 | NP_037413.1 | 2 | 1664 | ||
| NM_198391.2 | NP_938205.1 | 3 | 1858 | |||
| XM_005260682.4 | XP_005260739.1 | 3 | 2376 | |||
| XM_011529204.2 | XP_011527506.1 | 4 | 2513 | |||
| XM_011529205.2 | XP_011527507.1 | 4 | 2175 | |||
| GenBank mRNA | AB040902.1 | - | 2 | 1552 | ||
| AF169677.1 | - | 2 | 1561 | |||
| AK027297.1 | - | 2 | 1575 | |||
| AK027670.1 | - | 1 | 772 | |||
| AK027694.1 | - | 1 | 1293 | |||
| AK074883.1 | - | 2 | 1605 | |||
| AK074909.1 | - | 2 | 1533 | |||
| AY358319.1 | - | 1 | 1398 | |||
| BC020870.1 | - | 2 | 1571 | 4606852 | ||

FLRT3
Genomic Map