siRNA Details
| Gene Symbol: FJX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014344.3 | NP_055159.2 | 1 | 1504 | ||
| GenBank mRNA | AJ245599.1 | - | 1 | 1184 | ||
| AK026069.1 | - | 1 | 470 | |||
| AK315015.1 | - | 1 | 1273 | |||
| BC049171.1 | - | 1 | 1250 | 5482332 | ||
| BC071898.1 | - | 1 | 1250 | 5470218 | ||

FJX1
Genomic Map