siRNA Details
| Gene Symbol: ZNF318 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014345.2 | NP_055160.2 | 10 | 5614 | ||
| XM_011514451.2 | XP_011512753.1 | 10 | 5215 | |||
| GenBank mRNA | AF090114.1 | - | 10 | 5617 | ||
| AF121141.1 | - | 10 | 5352 | |||
| AK027061.1 | - | 1 | 193 | |||

ZNF318
Genomic Map