siRNA Details
| Gene Symbol: SOGA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB020696.1 | - | 1 | 250 | ||
| AK022023.1 | - | 1 | 440 | |||
| BC113405.1 | - | 1 | 200 | |||
| BC113433.1 | - | 1 | 200 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SOGA1| Gene Symbol: SOGA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB020696.1 | - | 1 | 250 | ||
| AK022023.1 | - | 1 | 440 | |||
| BC113405.1 | - | 1 | 200 | |||
| BC113433.1 | - | 1 | 200 | |||